The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033301.4 | NP_001028473.2 | 5 - 6 | 962 | 101 |
NM_001205355.1 | NP_001192284.1 | 5 - 6 | 1033 | 101 | |
GenBank mRNA | AK031946.1 | - | 4 - 5 | 438 | 101 |
AK145217.1 | - | 5 - 6 | 953 | 101 | |
AK162493.1 | - | 1 - 2 | 826 | 101 | |
AK173244.1 | - | 5 - 6 | 1033 | 101 | |
BC137632.1 | - | 5 - 6 | 1002 | 101 |