Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bap1
Entrez Gene ID
104416
Gene Name
Brca1 associated protein 1
Gene Aliases
2300006C11Rik, AA989761, AW553466, mKIAA0272, uch-x4
Chromosome Location
Chr.14: 31251455 - 31259930 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027088.2 NP_081364.1 8 - 9 786 63
GenBank mRNA AB047820.1 - 8 - 9 687 63
AK079374.1 - 8 - 9 802 63
AK129108.1 - 8 - 9 694 63
AK170576.1 - 8 - 9 787 63
AK171561.1 - 8 - 9 788 63
AK185783.1 - 2 - 3 171 63
AK189418.1 - 2 - 3 172 63
BC042489.1 - 8 - 9 737 63
BC050901.1 - 8 - 9 749 63

View Full Product Details