Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema3g
Entrez Gene ID
218877
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3G
Gene Aliases
AK129018
Chromosome Location
Chr.14: 31217873 - 31230352 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025379.1 NP_001020550.1 13 - 14 1550 112
GenBank mRNA AB127607.1 - 13 - 14 1550 112
AK129018.1 - 13 - 14 1565 112
AK137463.1 - 13 - 14 1567 112
BC115440.2 - 13 - 14 1549 112

View Full Product Details