Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr1
Entrez Gene ID
22388
Gene Name
WD repeat domain 1
Gene Aliases
Aip1, D5Wsu185e, rede
Chromosome Location
Chr.5: 38526813 - 38561800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011715.2 NP_035845.1 13 - 14 1627 90
GenBank mRNA AF020055.1 - 13 - 14 1627 90
AK004644.1 - 13 - 14 1735 90
AK004858.1 - 13 - 14 1698 90
AK151146.1 - 13 - 14 1624 90
AK153338.1 - 13 - 14 1638 90
AK165225.1 - 13 - 14 1597 90
AK165323.1 - 13 - 14 1598 90
AK167241.1 - 13 - 14 1653 90
AK167275.1 - 13 - 14 1695 90
AK167813.1 - 13 - 14 1625 90
AK169835.1 - 13 - 14 1721 90
AK169893.1 - 13 - 14 1657 90
AK170379.1 - 13 - 14 1637 90
AK207267.1 - 1 - 2 82 90
BC049117.1 - 13 - 14 1719 90
BC119358.1 - 13 - 14 1583 90
BC125295.1 - 13 - 14 1583 90

View Full Product Details