Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyn
Entrez Gene ID
17096
Gene Name
Yamaguchi sarcoma viral (v-yes-1) oncogene homolog
Gene Aliases
AA407514, Hck-2, p53Lyn, p56Lyn
Chromosome Location
Chr.4: 3678121 - 3791612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010747.2 NP_034877.2 2 - 3 324 146
GenBank mRNA AK028112.1 - 2 - 3 320 146
AK153038.1 - 2 - 3 321 146
BC003253.1 - 2 - 3 287 146
BC157998.1 - 2 - 3 156 146
M57697.1 - 2 - 3 160 146

View Full Product Details