The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026253.4 | NP_080529.1 | 3 - 4 | 1121 | 145 |
GenBank mRNA | AK014952.1 | - | 3 - 4 | 1086 | 145 |
AK015365.1 | - | 3 - 4 | 1125 | 145 | |
AK029909.1 | - | 3 - 4 | 1086 | 145 | |
AY775400.1 | - | 3 - 4 | 1190 | 145 |