Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ogdhl
Entrez Gene ID
239017
Gene Name
oxoglutarate dehydrogenase-like
Gene Aliases
-
Chromosome Location
Chr.14: 32322012 - 32348150 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081130.1 NP_001074599.1 23 - 24 3135 111
GenBank mRNA BC157970.1 - 22 - 23 2935 111
BC157971.1 - 22 - 23 2935 111

View Full Product Details