The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019944.2 | NP_064328.2 | 1 - 2 | 886 | 64 |
GenBank mRNA | BC132283.1 | - | 1 - 2 | 799 | 64 |
BC144718.1 | - | 1 - 2 | 800 | 64 |