Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrg1
Entrez Gene ID
546336
Gene Name
proline rich Gla (G-carboxyglutamic acid) 1
Gene Aliases
2010007L08Rik
Chromosome Location
Chr.X: 78449610 - 78583913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK008151.1 - 5 - 6 513 70

View Full Product Details