Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il17rb
Entrez Gene ID
50905
Gene Name
interleukin 17 receptor B
Gene Aliases
Evi27, IL-17ER, IL-17Rh1, IL17RH1, Il17br
Chromosome Location
Chr.14: 29995506 - 30008896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF208109.1 - 5 - 6 499 66

View Full Product Details