Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d1
Entrez Gene ID
57915
Gene Name
TBC1 domain family, member 1
Gene Aliases
1110062G02Rik, AI385682, AW555803, Tbc1, mKIAA1108
Chromosome Location
Chr.5: 64156310 - 64351486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289514.1 NP_001276443.1 6 - 7 1039 60
NM_019636.3 NP_062610.2 7 - 8 1620 60
GenBank mRNA AK122445.1 - 7 - 8 2429 60
AK135665.1 - 6 - 7 1210 60
AK135892.1 - 6 - 7 1304 60
AK148356.1 - 6 - 7 996 60
AK154591.1 - 8 - 9 1656 60
AK159256.1 - 7 - 8 1616 60
AK164923.1 - 3 - 4 324 60
BC059824.1 - 8 - 9 1985 60
BC117931.1 - 6 - 7 1288 60
U33005.1 - 7 - 8 1576 60

View Full Product Details