Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl3
Entrez Gene ID
50789
Gene Name
F-box and leucine-rich repeat protein 3
Gene Aliases
AU041772, AW212966, FBK, Fbl3a, Fbxl3a, Ovtm, Play68
Chromosome Location
Chr.14: 103079730 - 103099944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015822.2 NP_056637.1 2 - 3 700 80
GenBank mRNA AF176521.1 - 2 - 3 407 80
AK080863.1 - 2 - 3 709 80
AK142007.1 - 2 - 3 700 80
AK166128.1 - 2 - 3 581 80
AK167409.1 - 2 - 3 519 80
BC026835.1 - 1 - 2 366 80
BC037665.1 - 1 - 2 363 80
BC067203.1 - 2 - 3 582 80

View Full Product Details