Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema3d
Entrez Gene ID
108151
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D
Gene Aliases
4631426B19Rik
Chromosome Location
Chr.5: 12383166 - 12588943 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028882.4 NP_083158.3 15 - 16 2245 83
GenBank mRNA AK028900.1 - 15 - 16 2027 83
AK031704.1 - 15 - 16 1857 83
AK052671.1 - 15 - 16 2246 83
AK153839.1 - 15 - 16 1906 83
BC138131.1 - 15 - 16 1788 83

View Full Product Details