Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema3d
Entrez Gene ID
108151
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D
Gene Aliases
4631426B19Rik
Chromosome Location
Chr.5: 12383166 - 12588943 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028882.4 NP_083158.3 2 - 3 702 145
GenBank mRNA AK028900.1 - 2 - 3 484 145
AK051165.1 - 2 - 3 363 145
AK052671.1 - 2 - 3 703 145
AK087013.1 - 2 - 3 304 145
AK134412.1 - 2 - 3 338 145
AK153839.1 - 2 - 3 363 145
BC138131.1 - 2 - 3 245 145

View Full Product Details