Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcca
Entrez Gene ID
110821
Gene Name
propionyl-Coenzyme A carboxylase, alpha polypeptide
Gene Aliases
C79630
Chromosome Location
Chr.14: 122484414 - 122889945 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144844.2 NP_659093.2 21 - 22 1926 73
GenBank mRNA AK144814.1 - 4 - 5 329 73
AK148002.1 - 21 - 22 1916 73
AK183579.1 - 2 - 3 73
AK206880.1 - 2 - 3 82 73
AK208044.1 - 2 - 3 82 73
AK219004.1 - 2 - 3 82 73
AY046947.1 - 21 - 22 1943 73
BC006915.1 - 21 - 22 1923 73
BC049802.1 - 21 - 22 1892 73

View Full Product Details