Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slco5a1
- Entrez Gene ID
-
240726
- Gene Name
- solute carrier organic anion transporter family, member 5A1
- Gene Aliases
-
4732454B15, A630033C23Rik, AA407151
- Chromosome Location
-
Chr.1: 12866549 - 12992932 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_172841.2 |
NP_766429.2 |
6 - 7 |
2419 |
72 |
View Full Product Details