The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172841.2 | NP_766429.2 | 8 - 9 | 2726 | 80 |
GenBank mRNA | AV282837.2 | - | 2 - 3 | 95 | 80 |