Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rdh10
Entrez Gene ID
98711
Gene Name
retinol dehydrogenase 10 (all-trans)
Gene Aliases
3110069K09Rik, 4921506A21Rik, AI875664, AW549993, D1Ertd762e, m366Asp
Chromosome Location
Chr.1: 16105780 - 16133734 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133832.3 NP_598593.1 3 - 4 758 75
GenBank mRNA AF456767.1 - 3 - 4 866 75
AK149656.1 - 3 - 4 829 75
BC019796.1 - 3 - 4 758 75
BC038334.1 - 3 - 4 75
BC052546.1 - 3 - 4 75

View Full Product Details