Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyg2
Entrez Gene ID
332427
Gene Name
lysozyme G-like 2
Gene Aliases
Gm776
Chromosome Location
Chr.1: 37905583 - 37916493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033427.3 NP_001028599.1 1 - 2 260 82
GenBank mRNA AK132442.1 - 1 - 2 262 82
BC147562.1 - 1 - 2 197 82
BC147567.1 - 1 - 2 197 82

View Full Product Details