Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taf1
Entrez Gene ID
270627
Gene Name
TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
AU015687, B430306D02Rik, Ccg-1, Ccg1, KAT4, N-TAF1, TAFII250, Taf2a, p250
Chromosome Location
Chr.X: 101527574 - 101601789 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290729.1 NP_001277658.1 20 - 21 3195 102
GenBank mRNA AB299229.1 - 20 - 21 3122 102
AF022178.1 - 5 - 6 510 102
AK050691.1 - 5 - 6 566 102
BC047418.1 - 10 - 11 1308 102
BC051073.1 - 7 - 8 921 102
BC094568.1 - 2 - 3 479 102

View Full Product Details