Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zap70
Entrez Gene ID
22637
Gene Name
zeta-chain (TCR) associated protein kinase
Gene Aliases
Srk, ZAP-70, mrtle, mur
Chromosome Location
Chr.1: 36761798 - 36782821 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289765.1 NP_001276694.1 6 - 7 1033 68
NM_001289766.1 NP_001276695.1 6 - 7 998 68
NM_009539.3 NP_033565.2 6 - 7 990 68
GenBank mRNA AK039883.1 - 6 - 7 927 68
AK089056.1 - 6 - 7 963 68
BC029727.1 - 6 - 7 948 68
U04379.1 - 6 - 7 944 68
U77667.1 - 5 - 6 836 68

View Full Product Details