Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stx5a
Entrez Gene ID
56389
Gene Name
syntaxin 5A
Gene Aliases
0610031F24Rik, D19Ertd627e, Stx5
Chromosome Location
Chr.19: 8741424 - 8755642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167799.1 NP_001161271.1 5 - 6 597 117
NM_019829.4 NP_062803.4 4 - 5 896 117
GenBank mRNA AF232709.1 - 4 - 5 264 117
AK002722.1 - 5 - 6 579 117
AK166087.1 - 4 - 5 899 117
BC004849.1 - 5 - 6 530 117
BC021883.1 - 4 - 5 879 117
CN539678.1 - 5 - 6 561 117

View Full Product Details