Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx32
Entrez Gene ID
225861
Gene Name
sorting nexin 32
Gene Aliases
B930037P14Rik, Snx6b
Chromosome Location
Chr.19: 5495274 - 5510694 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024560.2 NP_001019731.1 6 - 7 721 62
GenBank mRNA AK037118.1 - - 712 62
AK143441.1 - 5 - 6 833 62
BC048922.1 - 6 - 7 721 62

View Full Product Details