The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168270.1 | NP_001161742.1 | 19 - 20 | 2008 | 85 |
NM_133794.2 | NP_598555.2 | 23 - 24 | 2354 | 85 | |
GenBank mRNA | AK030554.1 | - | 23 - 24 | 2301 | 85 |
AK088049.1 | - | 23 - 24 | 2301 | 85 | |
AK145632.1 | - | 23 - 24 | 2305 | 85 | |
AK155759.1 | - | 23 - 24 | 2303 | 85 | |
AK165454.1 | - | 23 - 24 | 2301 | 85 | |
AK170826.1 | - | 23 - 24 | 2303 | 85 | |
AK171293.1 | - | 23 - 24 | 2303 | 85 | |
BC018274.1 | - | 3 - 4 | 322 | 85 | |
BC023023.1 | - | 19 - 20 | 2008 | 85 | |
BC079854.1 | - | 23 - 24 | 2288 | 85 |