The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168270.1 | NP_001161742.1 | 6 - 7 | 613 | 73 |
NM_133794.2 | NP_598555.2 | 10 - 11 | 959 | 73 | |
GenBank mRNA | AK004814.1 | - | 10 - 11 | 893 | 73 |
AK030554.1 | - | 10 - 11 | 906 | 73 | |
AK088049.1 | - | 10 - 11 | 906 | 73 | |
AK145632.1 | - | 10 - 11 | 910 | 73 | |
AK155759.1 | - | 10 - 11 | 908 | 73 | |
AK165454.1 | - | 10 - 11 | 906 | 73 | |
AK167782.1 | - | 10 - 11 | 906 | 73 | |
AK170826.1 | - | 10 - 11 | 908 | 73 | |
AK171293.1 | - | 10 - 11 | 908 | 73 | |
BC023023.1 | - | 6 - 7 | 613 | 73 | |
BC079854.1 | - | 10 - 11 | 893 | 73 |