Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Twsg1
- Entrez Gene ID
-
65960
- Gene Name
- twisted gastrulation homolog 1 (Drosophila)
- Gene Aliases
-
1810013J15Rik, 9030422N06Rik, AW552143, D17Ertd403e, Tsg, Twg
- Chromosome Location
-
Chr.17: 65923065 - 65951187 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF292033.1 |
- |
- |
801 |
97 |
View Full Product Details