The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080129.2 | NP_001073598.1 | 5 - 6 | 1093 | 68 |
NM_001283048.1 | NP_001269977.1 | 5 - 6 | 776 | 68 | |
GenBank mRNA | AK011988.1 | - | 5 - 6 | 586 | 68 |
AK011992.1 | - | 5 - 6 | 586 | 68 | |
AK141845.1 | - | 5 - 6 | 1043 | 68 | |
AK146903.1 | - | 5 - 6 | 1060 | 68 | |
AK150305.1 | - | 5 - 6 | 1041 | 68 | |
AK150515.1 | - | 5 - 6 | 1028 | 68 | |
AK151936.1 | - | 5 - 6 | 1045 | 68 | |
AK152737.1 | - | 5 - 6 | 1045 | 68 | |
AK153281.1 | - | 5 - 6 | 1028 | 68 | |
AK159161.1 | - | 5 - 6 | 689 | 68 | |
AK165851.1 | - | 5 - 6 | 1050 | 68 | |
U39074.1 | - | 5 - 6 | 1024 | 68 |