Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd11
Entrez Gene ID
74007
Gene Name
BTB (POZ) domain containing 11
Gene Aliases
6330404E16Rik
Chromosome Location
Chr.10: 85386776 - 85660292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017525.1 NP_001017525.1 13 - 14 1664 81
NM_028709.2 NP_082985.2 15 - 16 3517 81
GenBank mRNA AK018115.1 - 5 - 6 978 81
AK039038.1 - 12 - 13 1460 81
AK044022.1 - 13 - 14 1555 81
BC059036.1 - 3 - 4 535 81
BC072592.1 - 15 - 16 81
BC096535.1 - 15 - 16 3517 81
BC141145.1 - 15 - 16 2589 81

View Full Product Details