The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161432.1 | NP_001154904.1 | 5 - 6 | 613 | 73 |
NM_001161433.1 | NP_001154905.1 | 5 - 6 | 568 | 73 | |
NM_175540.5 | NP_780749.3 | 5 - 6 | 568 | 73 | |
GenBank mRNA | AK034909.1 | - | 5 - 6 | 532 | 73 |
AK049134.1 | - | 5 - 6 | 575 | 73 | |
AK080072.1 | - | 5 - 6 | 532 | 73 | |
BC106845.1 | - | 5 - 6 | 554 | 73 |