Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ccny
- Entrez Gene ID
-
67974
- Gene Name
- cyclin Y
- Gene Aliases
-
1700025H17Rik, 3110050L10Rik, 4631402G10Rik, 5730405I09Rik
- Chromosome Location
-
Chr.18: 9312304 - 9450150 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC035524.1 |
- |
1 - 2 |
457 |
97 |
View Full Product Details