The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081189.1 | NP_001074658.1 | 3 - 4 | 686 | 98 |
GenBank mRNA | BC147845.1 | - | 3 - 4 | 544 | 98 |
BC147853.1 | - | 3 - 4 | 544 | 98 |