The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009205.2 | NP_033231.2 | 3 - 4 | 846 | 69 |
GenBank mRNA | AK052623.1 | - | 3 - 4 | 817 | 69 |
AK132275.1 | - | 3 - 4 | 818 | 69 | |
BC013441.1 | - | 3 - 4 | 788 | 69 |