Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esyt3
Entrez Gene ID
272636
Gene Name
extended synaptotagmin-like protein 3
Gene Aliases
D930024E11, D9Ertd280e, Fam62c, mKIAA4186
Chromosome Location
Chr.9: 99308739 - 99360405 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177775.3 NP_808443.2 2 - 3 512 82
GenBank mRNA AK086367.1 - 2 - 3 513 82
AK220532.1 - 2 - 3 500 82

View Full Product Details