The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026497.2 | NP_080773.1 | 6 - 7 | 1390 | 61 |
GenBank mRNA | AK002641.1 | - | 6 - 7 | 1391 | 61 |
AK083846.1 | - | 6 - 7 | 1348 | 61 | |
AK147867.1 | - | 6 - 7 | 1344 | 61 | |
BC050252.1 | - | 1 - 2 | 379 | 61 |