The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008020.3 | NP_032046.1 | 1 - 2 | 42 | 84 |
GenBank mRNA | AK144284.1 | - | 1 - 2 | 43 | 84 |