Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d15
Entrez Gene ID
66687
Gene Name
TBC1 domain family, member 15
Gene Aliases
4432405K22Rik, AU022358, AU023730, AW557774, Ly6dl, Rab7-GAP
Chromosome Location
Chr.10: 115197871 - 115251493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025706.3 NP_079982.3 13 - 14 1585 150
GenBank mRNA AK014477.1 - 13 - 14 1517 150
AK014795.1 - 8 - 9 968 150
AK078875.1 - 13 - 14 1531 150
AK135905.1 - 9 - 10 1061 150
AK147085.1 - 13 - 14 1548 150
AK162115.1 - 2 - 3 183 150
BC032087.1 - 13 - 14 1513 150
BC053395.1 - 13 - 14 1566 150
DQ054831.1 - 13 - 14 1516 150

View Full Product Details