Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d15
Entrez Gene ID
66687
Gene Name
TBC1 domain family, member 15
Gene Aliases
4432405K22Rik, AU022358, AU023730, AW557774, Ly6dl, Rab7-GAP
Chromosome Location
Chr.10: 115197871 - 115251493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025706.3 NP_079982.3 14 - 15 1686 166
GenBank mRNA AK014477.1 - 14 - 15 1618 166
AK014795.1 - 9 - 10 1069 166
AK078875.1 - 14 - 15 1632 166
AK135905.1 - 10 - 11 1162 166
AK147085.1 - 14 - 15 1649 166
AK162115.1 - 3 - 4 284 166
BC032087.1 - 14 - 15 1614 166
BC053395.1 - 14 - 15 1667 166
DQ054831.1 - 14 - 15 1617 166

View Full Product Details