Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fut4-ps1
Entrez Gene ID
14346
Gene Name
fucosyltransferase 4, pseudogene 1
Gene Aliases
-
Chromosome Location
Chr.17: 56734228 - 56739292 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033644.1 1 - 2 679 90
GenBank mRNA AK019884.1 - 1 - 2 679 90

View Full Product Details