Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tex16
Entrez Gene ID
83556
Gene Name
testis expressed gene 16
Gene Aliases
OTTMUSG00000018437
Chromosome Location
Chr.X: 111894858 - 112127326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031382.2 NP_113559.2 3 - 4 3386 101
GenBank mRNA AF285573.1 - 3 - 4 3386 101
BC132332.1 - 3 - 4 3410 101

View Full Product Details