The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145142.2 | NP_660124.2 | 1 - 2 | 268 | 65 |
GenBank mRNA | AK046415.1 | - | 1 - 2 | 120 | 65 |
AK047378.1 | - | 1 - 2 | 262 | 65 | |
AK169585.1 | - | 1 - 2 | 94 | 65 | |
AK169589.1 | - | 1 - 2 | 94 | 65 | |
AK171758.1 | - | 1 - 2 | 119 | 65 | |
BC056956.1 | - | 1 - 2 | 197 | 65 |