Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ascl4
Entrez Gene ID
67341
Gene Name
achaete-scute complex homolog 4 (Drosophila)
Gene Aliases
1700054F16Rik, bHLHa44
Chromosome Location
Chr.10: 85928491 - 85929647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK006781.1 - - 268 109
AK018959.1 - - 268 109

View Full Product Details