Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm2a
Entrez Gene ID
225876
Gene Name
lysine (K)-specific demethylase 2A
Gene Aliases
100043628, 5530401A10Rik, AA589516, AW536790, Cxxc8, Fbl11, Fbl7, Fbxl11, Gm4560, Jhdm1, Jhdm1a, lalina
Chromosome Location
Chr.19: 4316144 - 4398280 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001984.2 NP_001001984.2 1 - 2 766 152
GenBank mRNA AK037157.1 - 2 - 3 663 152
AK147028.1 - 1 - 2 758 152
AK155866.1 - 1 - 2 771 151
AK161980.1 - 1 - 2 767 152
AK169873.1 - 1 - 2 785 152
CB246224.1 - 2 - 3 605 152

View Full Product Details