Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntpcr
Entrez Gene ID
66566
Gene Name
nucleoside-triphosphatase, cancer-related
Gene Aliases
2310079N02Rik, AI449709
Chromosome Location
Chr.8: 125729976 - 125748235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025636.5 NP_079912.1 1 - 2 142 63
GenBank mRNA AK006433.1 - 1 - 2 50 63
AK010229.1 - 1 - 2 117 63
AK010823.1 - 1 - 2 143 63
AK019321.1 - 1 - 2 122 63
AK019421.1 - 1 - 2 113 63
AK086714.1 - 1 - 2 108 63
BC025168.1 - 1 - 2 50 63

View Full Product Details