The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028018.2 | NP_082294.1 | 9 - 10 | 1023 | 116 |
GenBank mRNA | AK010262.1 | - | 9 - 10 | 993 | 116 |
AK088192.1 | - | 9 - 10 | 978 | 116 | |
AK152830.1 | - | 9 - 10 | 990 | 116 | |
AK186831.1 | - | 3 - 4 | 269 | 116 | |
AK215347.1 | - | 3 - 4 | 253 | 116 | |
BC017605.1 | - | 9 - 10 | 989 | 116 |