Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb2
Entrez Gene ID
66557
Gene Name
BPI fold containing family B, member 2
Gene Aliases
2310034L21Rik, 2310069A01Rik, AV089125, Bpil1, LPLUNC2, RYSR
Chromosome Location
Chr.2: 153875045 - 153895270 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025631.3 NP_079907.2 2 - 3 219 101
GenBank mRNA AK009346.1 - 2 - 3 192 101
AK009504.1 - 2 - 3 194 101
AK009619.1 - 2 - 3 192 101
AK009776.1 - 2 - 3 176 101
AK009794.1 - 2 - 3 190 101
AK009871.1 - 2 - 3 194 101
AK010130.1 - 2 - 3 176 101
AK010154.1 - 2 - 3 190 101
AK010157.1 - 2 - 3 220 101
AK010215.1 - 2 - 3 190 101
AK028156.1 - 2 - 3 190 101
AK028158.1 - 2 - 3 193 101
AK075874.1 - 2 - 3 190 101
AK160516.1 - 2 - 3 190 101
BC116681.1 - 2 - 3 174 101
BC116727.1 - 2 - 3 174 101

View Full Product Details