Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpifb3
Entrez Gene ID
378700
Gene Name
BPI fold containing family B, member 3
Gene Aliases
Lplunc3, Rya3
Chromosome Location
Chr.2: 153918230 - 153932996 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194357.1 NP_919338.2 7 - 8 777 83
GenBank mRNA AK132627.1 - 2 - 3 129 83
BC048084.1 - 7 - 8 777 83

View Full Product Details