Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca8a
Entrez Gene ID
217258
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8a
Gene Aliases
-
Chromosome Location
Chr.11: 110025634 - 110097583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153145.4 NP_694785.3 26 - 27 3629 77
GenBank mRNA AB255881.1 - 25 - 26 3548 77
AB255882.1 - 26 - 27 3636 77
AF498360.1 - 25 - 26 3557 77
AK031843.1 - 7 - 8 860 77
AY732492.1 - 25 - 26 3557 77
BC026496.1 - 6 - 7 754 77
BC060032.1 - 8 - 9 1123 77

View Full Product Details