Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca8a
Entrez Gene ID
217258
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8a
Gene Aliases
-
Chromosome Location
Chr.11: 110025634 - 110097583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153145.4 NP_694785.3 3 - 4 377 127
GenBank mRNA AB255881.1 - 2 - 3 296 127
AB255882.1 - 3 - 4 384 127
AF498360.1 - 2 - 3 302 127
AY732492.1 - 2 - 3 302 127

View Full Product Details