Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp1a
Entrez Gene ID
195046
Gene Name
NLR family, pyrin domain containing 1A
Gene Aliases
CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044
Chromosome Location
Chr.11: 71091197 - 71144704 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004142.2 NP_001004142.2 11 - 12 3288 74
GenBank mRNA AY355339.1 - 12 - 13 3373 74
DQ117601.1 - 10 - 11 3229 74
KC539856.1 - 10 - 11 3070 74
KC539857.1 - 10 - 11 3070 74
KC539858.1 - 10 - 11 3070 74
KC539860.1 - 10 - 11 3070 74
KC539861.1 - 10 - 11 3070 74

View Full Product Details