Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxw24
Entrez Gene ID
382106
Gene Name
F-box and WD-40 domain protein 24
Gene Aliases
EG382106, Gm5162
Chromosome Location
Chr.9: 109601116 - 109626059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013776.4 NP_001013798.2 3 - 4 323 84
GenBank mRNA BC080701.1 - 3 - 4 324 84
CF913977.1 - 3 - 4 308 84
CJ075113.1 - 3 - 4 323 84
GQ418172.1 - 3 - 4 324 84

View Full Product Details